Product Details

SNP ID
rs201305704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22895412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACCCCACTGTCACTCTGTTCCC[A/G]CCCTCCTCTGAGGAGCTCCAAGCCA
Phenotype
MIM: 147220
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGLC1 PubMed Links

Gene Details

Gene
IGLC1
Gene Name
immunoglobulin lambda constant 1
There are no transcripts associated with this gene.

Gene
IGLJ3
Gene Name
immunoglobulin lambda joining 3
There are no transcripts associated with this gene.

Gene
IGLL5
Gene Name
immunoglobulin lambda like polypeptide 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178126.1 637 Silent Mutation CCA,CCG P121P NP_001171597.1
NM_001256296.1 637 Silent Mutation CCA,CCG P46P NP_001243225.1
Gene
MIR5571
Gene Name
microRNA 5571
There are no transcripts associated with this gene.

View Full Product Details