Product Details

SNP ID
rs199675711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50264962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGCACCTCACTGCTCAATCT[C/T]CAGGCTGCCAGGTGGCTTCGGTGGC
Phenotype
MIM: 602898 MIM: 602399
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPK11 PubMed Links

Gene Details

Gene
MAPK11
Gene Name
mitogen-activated protein kinase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002751.6 906 Missense Mutation AAG,GAG K361E NP_002742.3
XM_011530706.1 906 Missense Mutation AAG,GAG K253E XP_011529008.1
Gene
MAPK12
Gene Name
mitogen-activated protein kinase 12
There are no transcripts associated with this gene.

View Full Product Details