Product Details

SNP ID
rs201377055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44883178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCCAGCAGTGGGGCAGGGCTAG[G/T]GGTCGTCATGGTGACCTGGAGCAGC
Phenotype
MIM: 616727
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PHF21B PubMed Links

Gene Details

Gene
PHF21B
Gene Name
PHD finger protein 21B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135862.2 1655 Missense Mutation ACT,CCT T460P NP_001129334.1
NM_001242450.1 1655 Missense Mutation ACT,CCT T448P NP_001229379.1
NM_001284296.1 1655 Missense Mutation ACT,CCT T298P NP_001271225.1
NM_138415.4 1655 Missense Mutation ACT,CCT T502P NP_612424.1
XM_006724122.1 1655 Missense Mutation ACT,CCT T460P XP_006724185.1
XM_011529855.1 1655 Intron XP_011528157.1

View Full Product Details