Product Details

SNP ID
rs201445071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27217966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGTGCTCACCTATCCTCTGATC[C/G]GCAACTATGGCATCCCCCCAGATGA
Phenotype
MIM: 114010 MIM: 604024
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATRAID PubMed Links

Gene Details

Gene
ATRAID
Gene Name
all-trans retinoic acid induced differentiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170795.1 334 Intron NP_001164266.1
NM_016085.4 334 Intron NP_057169.2
NM_080592.3 334 Intron NP_542159.3
Gene
CAD
Gene Name
carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306079.1 334 Missense Mutation CGC,GGC R58G NP_001293008.1
NM_004341.4 334 Missense Mutation CGC,GGC R58G NP_004332.2
XM_005264555.3 334 Missense Mutation CGC,GGC R58G XP_005264612.1
XM_006712101.2 334 Missense Mutation CGC,GGC R58G XP_006712164.1
Gene
SLC5A6
Gene Name
solute carrier family 5 member 6
There are no transcripts associated with this gene.

View Full Product Details