Product Details

SNP ID
rs201216936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27366862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCTTCCTTCCAGCCATGGCCGG[C/T]TGTCCACCACTACCACCCGAAACCG
Phenotype
MIM: 606687 MIM: 604883 MIM: 605963
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2B4 PubMed Links

Gene Details

Gene
EIF2B4
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034116.1 1228 Missense Mutation NP_001029288.1
NM_001318965.1 1228 Missense Mutation NP_001305894.1
NM_001318966.1 1228 Missense Mutation NP_001305895.1
NM_001318967.1 1228 Missense Mutation NP_001305896.1
NM_001318968.1 1228 Missense Mutation NP_001305897.1
NM_001318969.1 1228 Missense Mutation NP_001305898.1
NM_015636.3 1228 Missense Mutation NP_056451.3
NM_172195.3 1228 Missense Mutation NP_751945.2
XM_006712132.1 1228 Missense Mutation XP_006712195.1
XM_011533147.2 1228 Missense Mutation XP_011531449.1
Gene
GTF3C2
Gene Name
general transcription factor IIIC subunit 2
There are no transcripts associated with this gene.

Gene
LOC105374363
Gene Name
uncharacterized LOC105374363
There are no transcripts associated with this gene.

Gene
SNX17
Gene Name
sorting nexin 17
There are no transcripts associated with this gene.

View Full Product Details