Product Details
- SNP ID
-
rs201346228
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:171435140 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATTCCAGGTCACCTATAGCCTAT[A/G]AGAGAGGAAGAATATATTTTGACAA
- Phenotype
-
MIM: 612515
MIM: 609525
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DCAF17
PubMed Links
Gene Details
- Gene
- DCAF17
- Gene Name
- DDB1 and CUL4 associated factor 17
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001164821.1 |
163 |
Missense Mutation |
AAG,GAG |
K62E |
NP_001158293.1 |
NM_025000.3 |
163 |
Missense Mutation |
AAG,GAG |
K62E |
NP_079276.2 |
XM_006712767.1 |
163 |
UTR 5 |
|
|
XP_006712830.1 |
XM_006712768.1 |
163 |
UTR 5 |
|
|
XP_006712831.1 |
XM_011511881.1 |
163 |
Missense Mutation |
AAG,GAG |
K62E |
XP_011510183.1 |
XM_011511882.1 |
163 |
Missense Mutation |
AAG,GAG |
K62E |
XP_011510184.1 |
XM_017004995.1 |
163 |
Missense Mutation |
AAG,GAG |
K62E |
XP_016860484.1 |
XM_017004996.1 |
163 |
Missense Mutation |
AAG,GAG |
K62E |
XP_016860485.1 |
XM_017004997.1 |
163 |
Missense Mutation |
AAG,GAG |
K62E |
XP_016860486.1 |
XM_017004998.1 |
163 |
UTR 5 |
|
|
XP_016860487.1 |
XM_017004999.1 |
163 |
Missense Mutation |
AAG,GAG |
K62E |
XP_016860488.1 |
XM_017005000.1 |
163 |
Missense Mutation |
AAG,GAG |
K62E |
XP_016860489.1 |
XM_017005001.1 |
163 |
Missense Mutation |
AAG,GAG |
K62E |
XP_016860490.1 |
XM_017005002.1 |
163 |
UTR 5 |
|
|
XP_016860491.1 |
- Gene
- METTL8
- Gene Name
- methyltransferase like 8
View Full Product Details