Product Details

SNP ID
rs201383709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:30234471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACACCCAGCCCATGGAGGAGAT[C/T]GGCCTCAGCCCCCGCAAGGATGGCC
Phenotype
MIM: 611763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LBH PubMed Links

Gene Details

Gene
LBH
Gene Name
limb bud and heart development
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030915.3 301 Silent Mutation ATC,ATT I31I NP_112177.2
XM_017005059.1 301 Silent Mutation ATC,ATT I148I XP_016860548.1

View Full Product Details