Product Details

SNP ID
rs201363015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219283178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCAGTCCTGGGGCTGGTGCTT[G/T]TCGCTCTGTTTCTACATCTACCACC
Phenotype
MIM: 604139 MIM: 601773
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB2 PubMed Links

Gene Details

Gene
DNAJB2
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039550.1 728 Missense Mutation TGT,TTT C164F NP_001034639.1
NM_006736.5 728 Missense Mutation TGT,TTT C164F NP_006727.2
Gene
PTPRN
Gene Name
protein tyrosine phosphatase, receptor type N
There are no transcripts associated with this gene.

View Full Product Details