Product Details

SNP ID
rs200876442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166198889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATCATCTCTGTCTCCATCTTTT[A/G]TGTATATACTTGATATATTTTTGAC
Phenotype
MIM: 603415
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 6025 Missense Mutation ACA,ATA T1906I NP_002968.1
XM_005246757.2 6025 Missense Mutation ACA,ATA T1917I XP_005246814.1
XM_011511616.2 6025 Missense Mutation ACA,ATA T1917I XP_011509918.1
XM_011511617.2 6025 Missense Mutation ACA,ATA T1917I XP_011509919.1
XM_011511618.2 6025 Missense Mutation ACA,ATA T1906I XP_011509920.1
XM_011511619.2 6025 Intron XP_011509921.1
XM_017004668.1 6025 Missense Mutation ACA,ATA T1788I XP_016860157.1
XM_017004669.1 6025 Missense Mutation ACA,ATA T1669I XP_016860158.1

View Full Product Details