Product Details

SNP ID
rs200935158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219182036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTGGACCAGCAGAGCCTGCCA[A/G]GTGAACCAGAGGAGACCCTAAGCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPPD1 PubMed Links

Gene Details

Gene
CNPPD1
Gene Name
cyclin Pas1/PHO80 domain containing 1
There are no transcripts associated with this gene.

Gene
FAM134A
Gene Name
family with sequence similarity 134 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321109.1 1202 Missense Mutation AGT,GGT S140G NP_001308038.1
NM_001321110.1 1202 Missense Mutation AGT,GGT S140G NP_001308039.1
NM_024293.5 1202 Missense Mutation AGT,GGT S347G NP_077269.3
XM_005246848.2 1202 Missense Mutation AGT,GGT S347G XP_005246905.1

View Full Product Details