Product Details

SNP ID
rs201034493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130181591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCGCCATTCCGCCACGGCGCCG[A/G]AAGTCGTCATCAAGCTGCGCGCAGA
Phenotype
MIM: 613450 MIM: 610457
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MZT2B PubMed Links

Gene Details

Gene
MZT2B
Gene Name
mitotic spindle organizing protein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263795.4 Intron XP_005263852.1
XM_011511886.2 Intron XP_011510188.1
XM_011511887.2 Intron XP_011510189.1
XM_011511888.2 Intron XP_011510190.1
XM_011511889.2 Intron XP_011510191.1
XM_011511890.2 Intron XP_011510192.1
Gene
SMPD4
Gene Name
sphingomyelin phosphodiesterase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171083.2 Intron NP_001164554.1
NM_017751.4 Intron NP_060221.2
NM_017951.4 Intron NP_060421.2
XM_011511443.2 Intron XP_011509745.1
XM_011511444.2 Intron XP_011509746.1
XM_011511445.2 Intron XP_011509747.1
XM_017004448.1 Intron XP_016859937.1
XM_017004449.1 Intron XP_016859938.1

View Full Product Details