Product Details

SNP ID
rs201115978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:199933153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCCCTATATTCCTCTGGTAACT[C/T]GGCCAGTGTTTTCAAGGCTCTGTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf69 PubMed Links

Gene Details

Gene
C2orf69
Gene Name
chromosome 2 open reading frame 69
There are no transcripts associated with this gene.

Gene
TYW5
Gene Name
tRNA-yW synthesizing protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039693.2 1128 Missense Mutation AAG,GAG K288E NP_001034782.1
XM_017003310.1 1128 Missense Mutation AAG,GAG K183E XP_016858799.1

View Full Product Details