Product Details

SNP ID
rs200430731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241678496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCATGAGCTGGTGGAAACACC[A/G]GAGCGCCCGCTCCTGGAAAGCCCCG
Phenotype
MIM: 611338 MIM: 188345 MIM: 608525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG4B PubMed Links

Gene Details

Gene
ATG4B
Gene Name
autophagy related 4B cysteine peptidase
There are no transcripts associated with this gene.

Gene
DTYMK
Gene Name
deoxythymidylate kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165031.1 506 Missense Mutation CGG,TGG R138W NP_001158503.1
NM_001320902.1 506 Missense Mutation CGG,TGG R119W NP_001307831.1
NM_001320903.1 506 Silent Mutation TCC,TCT S107S NP_001307832.1
NM_001320904.1 506 Missense Mutation CGG,TGG R143W NP_001307833.1
NM_001320905.1 506 Missense Mutation CGG,TGG R201W NP_001307834.1
NM_012145.3 506 Missense Mutation CGG,TGG R162W NP_036277.2
Gene
ING5
Gene Name
inhibitor of growth family member 5
There are no transcripts associated with this gene.

View Full Product Details