Product Details

SNP ID
rs200376025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9585308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTAGTTTTCAGCCCCTTCTGCC[A/G]CATCACATTCTTCTCCTGCACTGTC
Phenotype
MIM: 609009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YWHAQ PubMed Links

Gene Details

Gene
YWHAQ
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006826.3 913 Missense Mutation NP_006817.1

View Full Product Details