Product Details

SNP ID
rs200193900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27443109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGCACTAGCTCTGGTGAACCT[A/G]CTCTTGTCCGTTGCCTGCTCCCTGG
Phenotype
MIM: 607386 MIM: 606010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT172 PubMed Links

Gene Details

Gene
IFT172
Gene Name
intraflagellar transport 172
There are no transcripts associated with this gene.

Gene
KRTCAP3
Gene Name
keratinocyte associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168364.1 356 Silent Mutation CTA,CTG L103L NP_001161836.1
NM_001321325.1 356 Silent Mutation CTA,CTG L103L NP_001308254.1
NM_173853.3 356 Silent Mutation CTA,CTG L103L NP_776252.2
Gene
NRBP1
Gene Name
nuclear receptor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321357.1 356 Intron NP_001308286.1
NM_001321358.1 356 Intron NP_001308287.1
NM_001321359.1 356 Intron NP_001308288.1
NM_001321361.1 356 Intron NP_001308290.1
NM_001321362.1 356 Intron NP_001308291.1
NM_001321363.1 356 Intron NP_001308292.1
NM_013392.3 356 Intron NP_037524.1

View Full Product Details