Product Details

SNP ID
rs200405610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95180185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCAGTTCCTCAACCTGATGTG[A/G]TTTTCCAATTGAAGAGAGGGGACAA
Phenotype
MIM: 194500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF2 PubMed Links

Gene Details

Gene
ZNF2
Gene Name
zinc finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017396.2 670 Missense Mutation ATT,GTT I21V NP_001017396.1
NM_001282398.1 670 Intron NP_001269327.1
NM_001291604.1 670 Intron NP_001278533.1
NM_001291605.1 670 Missense Mutation ATT,GTT I63V NP_001278534.1
NM_021088.3 670 Missense Mutation ATT,GTT I63V NP_066574.2

View Full Product Details