Product Details

SNP ID
rs200336628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:172088122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCTGTCTGCTGGCTCCCCACC[A/C]GTGCCGCCCGGCTGGAACCCTAACT
Phenotype
MIM: 600029 MIM: 610267
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DLX1 PubMed Links

Gene Details

Gene
DLX1
Gene Name
distal-less homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038493.1 831 UTR 3 NP_001033582.1
NM_178120.4 831 Silent Mutation CCA,CCC P211P NP_835221.2
Gene
METAP1D
Gene Name
methionyl aminopeptidase type 1D (mitochondrial)
There are no transcripts associated with this gene.

View Full Product Details