Product Details

SNP ID
rs199739977
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:181458197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACGTGCACGTGTCTCGCTTTAGG[C/G]TCCTAGTGGGTGCGCCCACTGCCAA
Phenotype
MIM: 192975
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ITGA4 PubMed Links
Additional Information
For this assay, SNP(s) [rs539227893] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ITGA4
Gene Name
integrin subunit alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000885.5 962 Missense Mutation CTC,GTC L67V NP_000876.3
NM_001316312.1 962 Missense Mutation CTC,GTC L67V NP_001303241.1

View Full Product Details