Product Details
- SNP ID
-
rs199782343
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:219055450 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACCAGTGTCCCATGCTTTGTGAG[C/T]GGGGCGTAGGCCCCGAGGGCCACGT
- Phenotype
-
MIM: 600726
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IHH
PubMed Links
Gene Details
- Gene
- IHH
- Gene Name
- indian hedgehog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002181.3 |
1042 |
Silent Mutation |
CCA,CCG |
P331P |
NP_002172.2 |
- Gene
- MIR3131
- Gene Name
- microRNA 3131
There are no transcripts associated with this gene.
View Full Product Details