Product Details
- SNP ID
-
rs200455799
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:222571938 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGGTCAGCTCAAATTTGGTGATA[A/G]CGTCAGGATGAAGGACCCCAAGCTT
- Phenotype
-
MIM: 609690
MIM: 612827
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FARSB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7185] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FARSB
- Gene Name
- phenylalanyl-tRNA synthetase beta subunit
- Gene
- SGPP2
- Gene Name
- sphingosine-1-phosphate phosphatase 2
There are no transcripts associated with this gene.
View Full Product Details