Product Details

SNP ID
rs201614590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38666178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTCCCTATGGCTTCGGTGACCA[A/G]GGCCGTGTTTGGAGAGCTGCCCTCG
Phenotype
MIM: 137030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALM PubMed Links

Gene Details

Gene
GALM
Gene Name
galactose mutarotase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138801.2 269 Missense Mutation AAG,AGG K6R NP_620156.1
XM_011532540.1 269 Missense Mutation AAG,AGG K6R XP_011530842.1

View Full Product Details