Product Details

SNP ID
rs201642678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24183307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTTACAGTCCCACCATTTGTTG[A/C]TCCTCTGTTTCAAAGACAGCAAGAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM228A PubMed Links

Gene Details

Gene
FAM228A
Gene Name
family with sequence similarity 228 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040710.2 331 Missense Mutation GAT,GCT D62A NP_001035800.1

View Full Product Details