Product Details

SNP ID
rs201698250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86751916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGAATTTTGATTCTGACATTAG[C/T]AGTGTGGGAATAGATGGCTGCTGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RMND5A PubMed Links

Gene Details

Gene
RMND5A
Gene Name
required for meiotic nuclear division 5 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022780.3 683 Silent Mutation AGC,AGT S102S NP_073617.1

View Full Product Details