Product Details

SNP ID
rs202198683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:157415768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGACACTTCCCTTTCTGCTCTTC[C/T]GGGGCAGGGTCCCAAACATGCTTGA
Phenotype
MIM: 604448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYTIP PubMed Links

Gene Details

Gene
CYTIP
Gene Name
cytohesin 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004288.4 1252 Missense Mutation NP_004279.3
XM_017005386.1 1252 Missense Mutation XP_016860875.1

View Full Product Details