Product Details

SNP ID
rs201633531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74426516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTGGGGGTGTCTCCAGCCTTGC[A/G]CCCTCCCGCTGGGTCAGGATGTCTG
Phenotype
MIM: 602288
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf81 PubMed Links

Gene Details

Gene
C2orf81
Gene Name
chromosome 2 open reading frame 81
There are no transcripts associated with this gene.

Gene
RTKN
Gene Name
rhotekin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015055.1 1921 Silent Mutation GGC,GGT G473G NP_001015055.1
NM_001015056.1 1921 Silent Mutation GGC,GGT G423G NP_001015056.1
NM_033046.2 1921 Silent Mutation GGC,GGT G460G NP_149035.1
XM_005264478.2 1921 UTR 3 XP_005264535.1
XM_005264479.4 1921 UTR 3 XP_005264536.1
XM_017004635.1 1921 Silent Mutation GGC,GGT G460G XP_016860124.1
XM_017004636.1 1921 UTR 3 XP_016860125.1
XM_017004637.1 1921 UTR 3 XP_016860126.1
XM_017004638.1 1921 Silent Mutation GGC,GGT G343G XP_016860127.1
XM_017004639.1 1921 Intron XP_016860128.1
XM_017004640.1 1921 UTR 3 XP_016860129.1
Gene
WDR54
Gene Name
WD repeat domain 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320823.1 1921 Intron NP_001307752.1
NM_001320824.1 1921 Intron NP_001307753.1
NM_001320825.1 1921 Intron NP_001307754.1
NM_032118.3 1921 Intron NP_115494.1
XM_017005064.1 1921 Intron XP_016860553.1

View Full Product Details