Product Details

SNP ID
rs201730025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32175357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTACCTGACCATTTTTGATCTT[C/T]TTAGGTAAGTTTTTAGGAAATCTTA
Phenotype
MIM: 611148
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC30A6 PubMed Links

Gene Details

Gene
SLC30A6
Gene Name
solute carrier family 30 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006712048.3 266 Intron XP_006712111.1
XM_011532959.2 266 Missense Mutation CTT,TTT L72F XP_011531261.1
XM_011532960.2 266 Missense Mutation CTT,TTT L23F XP_011531262.1
XM_011532961.2 266 Intron XP_011531263.1
XM_011532962.2 266 Intron XP_011531264.1
XM_011532963.2 266 Missense Mutation CTT,TTT L72F XP_011531265.1
XM_017004462.1 266 Intron XP_016859951.1
XM_017004463.1 266 Missense Mutation CTT,TTT L72F XP_016859952.1
XM_017004464.1 266 Missense Mutation CTT,TTT L72F XP_016859953.1
XM_017004465.1 266 Missense Mutation CTT,TTT L72F XP_016859954.1
XM_017004466.1 266 Missense Mutation CTT,TTT L72F XP_016859955.1

View Full Product Details