Product Details

SNP ID
rs201844790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24183561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGCTGCATGCCAGCTCGCCCT[A/G]CTTCACTTTCACTTCACACTGTGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM228A PubMed Links

Gene Details

Gene
FAM228A
Gene Name
family with sequence similarity 228 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040710.2 463 Missense Mutation TAC,TGC Y106C NP_001035800.1

View Full Product Details