Product Details

SNP ID
rs201889290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43832024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTTCACATACACCTCCCCCAGGA[A/G]GGTCCCCGCGCGCCCCAGCCTCCCG
Phenotype
MIM: 605459 MIM: 605460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCG5 PubMed Links

Gene Details

Gene
ABCG5
Gene Name
ATP binding cassette subfamily G member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022436.2 83 Missense Mutation CTC,TTC L109F NP_071881.1
XM_005264480.3 83 Missense Mutation CTC,TTC L109F XP_005264537.1
XM_006712073.3 83 Missense Mutation CTC,TTC L109F XP_006712136.1
XM_006712074.3 83 Missense Mutation CTC,TTC L109F XP_006712137.1
XM_011533024.2 83 Missense Mutation CTC,TTC L109F XP_011531326.1
XM_011533025.2 83 Missense Mutation CTC,TTC L28F XP_011531327.1
XM_011533026.2 83 Missense Mutation CTC,TTC L109F XP_011531328.1
XM_011533027.2 83 Intron XP_011531329.1
XM_011533028.2 83 Intron XP_011531330.1
Gene
ABCG8
Gene Name
ATP binding cassette subfamily G member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022437.2 83 Intron NP_071882.1
XM_005264483.3 83 Intron XP_005264540.1
XM_011533029.2 83 UTR 5 XP_011531331.1
XM_011533030.2 83 Intron XP_011531332.1
Gene
DYNC2LI1
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
There are no transcripts associated with this gene.

View Full Product Details