Product Details

SNP ID
rs201912686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50191806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGAGGACGCTGAGAAGGATGC[C/T]CGAACCGTGAAGCTGCTGCTTCTGG
Phenotype
MIM: 139330 MIM: 601124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNAT1 PubMed Links

Gene Details

Gene
GNAT1
Gene Name
G protein subunit alpha transducin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000172.3 197 Silent Mutation GCC,GCT A27A NP_000163.2
NM_144499.2 197 Silent Mutation GCC,GCT A27A NP_653082.1
Gene
SEMA3F
Gene Name
semaphorin 3F
There are no transcripts associated with this gene.

View Full Product Details