Product Details
- SNP ID
-
rs200883805
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:161499242 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTGTTGGGCCCCAAGGCCCTAGA[G/T]GCTACAAAGGAGAGAAAGGTAGGTA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OTOL1
PubMed Links
Gene Details
- Gene
- OTOL1
- Gene Name
- otolin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080440.1 |
436 |
Missense Mutation |
GGC,TGC |
G146C |
NP_001073909.1 |
View Full Product Details