Product Details

SNP ID
rs201026350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:152340618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAGTTTGGAAACTATCAGCAGG[C/T]ACCTGCAACTAAACCCACAAGCGTA
Phenotype
MIM: 606516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBNL1 PubMed Links

Gene Details

Gene
MBNL1
Gene Name
muscleblind like splicing regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314057.1 Intron NP_001300986.1
NM_021038.4 Intron NP_066368.2
NM_207292.2 Intron NP_997175.1
NM_207293.1 Intron NP_997176.1
NM_207294.1 Intron NP_997177.1
NM_207295.1 Intron NP_997178.1
NM_207296.1 Intron NP_997179.1
NM_207297.1 Intron NP_997180.1
XM_005247457.4 Intron XP_005247514.1
XM_005247458.4 Intron XP_005247515.1
XM_005247460.4 Intron XP_005247517.1
XM_005247461.4 Intron XP_005247518.1
XM_005247462.4 Intron XP_005247519.1
XM_005247463.4 Intron XP_005247520.1
XM_005247464.4 Intron XP_005247521.1
XM_005247465.4 Intron XP_005247522.1
XM_005247466.4 Intron XP_005247523.1
XM_005247467.4 Intron XP_005247524.1
XM_005247468.4 Intron XP_005247525.1
XM_005247471.4 Intron XP_005247528.1
XM_005247474.3 Intron XP_005247531.1
XM_005247475.3 Intron XP_005247532.1
XM_005247476.4 Intron XP_005247533.1
XM_005247477.4 Intron XP_005247534.1
XM_006713639.1 Intron XP_006713702.1
XM_011512843.2 Intron XP_011511145.1
XM_011512845.2 Intron XP_011511147.1
XM_011512846.2 Intron XP_011511148.1
XM_011512847.2 Intron XP_011511149.1
XM_011512848.2 Intron XP_011511150.1
XM_011512849.2 Intron XP_011511151.1
XM_017006423.1 Intron XP_016861912.1
XM_017006424.1 Intron XP_016861913.1
XM_017006425.1 Intron XP_016861914.1
XM_017006426.1 Intron XP_016861915.1
XM_017006427.1 Intron XP_016861916.1
XM_017006428.1 Intron XP_016861917.1
XM_017006429.1 Intron XP_016861918.1
XM_017006430.1 Intron XP_016861919.1
XM_017006431.1 Intron XP_016861920.1
XM_017006432.1 Intron XP_016861921.1
XM_017006433.1 Intron XP_016861922.1
XM_017006434.1 Intron XP_016861923.1
XM_017006435.1 Intron XP_016861924.1
XM_017006436.1 Intron XP_016861925.1
XM_017006437.1 Intron XP_016861926.1
XM_017006438.1 Intron XP_016861927.1
XM_017006439.1 Intron XP_016861928.1
XM_017006440.1 Intron XP_016861929.1
XM_017006441.1 Intron XP_016861930.1
XM_017006442.1 Intron XP_016861931.1
XM_017006443.1 Intron XP_016861932.1
Gene
TMEM14EP
Gene Name
transmembrane protein 14E, pseudogene
There are no transcripts associated with this gene.

View Full Product Details