Product Details

SNP ID
rs201114014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:19951060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAATTTCAAGAGAGTACCATTGG[G/T]GGTGAGATTTTCTTTTTTCCCTGCT
Phenotype
MIM: 179512
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EFHB PubMed Links

Gene Details

Gene
EFHB
Gene Name
EF-hand domain family member B
There are no transcripts associated with this gene.

Gene
RAB5A
Gene Name
RAB5A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292048.1 697 Silent Mutation GGG,GGT G54G NP_001278977.1
NM_004162.4 697 Silent Mutation GGG,GGT G54G NP_004153.2

View Full Product Details