Product Details

SNP ID
rs200743162
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98283169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGGTTAATTTCTTGGCCAAAAA[C/T]AGGATGATATCTCTGTCTGAATGCA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR5H2 PubMed Links

Gene Details

Gene
OR5H2
Gene Name
olfactory receptor family 5 subfamily H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005482.1 282 Silent Mutation AAC,AAT N94N NP_001005482.1

View Full Product Details