Product Details
- SNP ID
-
rs201412251
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:161496943 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAGAGATGCCAAAGGGTCTAAAG[C/T]CATCCAGTGGCCCACCTCCAGAAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OTOL1
PubMed Links
Gene Details
- Gene
- OTOL1
- Gene Name
- otolin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080440.1 |
136 |
Missense Mutation |
CCA,TCA |
P46S |
NP_001073909.1 |
View Full Product Details