Product Details

SNP ID
rs200829478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169778882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTATTGCTAATCCTTCTTCTAC[A/G]GAGATATCTAGTATTACTGGAAACA
Phenotype
MIM: 608534 MIM: 606042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTRT3 PubMed Links

Gene Details

Gene
ACTRT3
Gene Name
actin related protein T3
There are no transcripts associated with this gene.

Gene
MYNN
Gene Name
myoneurin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185118.1 1568 Silent Mutation ACA,ACG T127T NP_001172047.1
NM_001185119.1 1568 Silent Mutation ACA,ACG T127T NP_001172048.1
NM_018657.4 1568 Silent Mutation ACA,ACG T127T NP_061127.1
XM_005247621.4 1568 Silent Mutation ACA,ACG T127T XP_005247678.1
XM_005247622.3 1568 Intron XP_005247679.1
XM_005247624.3 1568 Silent Mutation ACA,ACG T13T XP_005247681.1
XM_011512987.1 1568 Silent Mutation ACA,ACG T127T XP_011511289.1
XM_017006864.1 1568 Silent Mutation ACA,ACG T127T XP_016862353.1
XM_017006865.1 1568 Silent Mutation ACA,ACG T13T XP_016862354.1
XM_017006866.1 1568 Silent Mutation ACA,ACG T13T XP_016862355.1
XM_017006867.1 1568 Intron XP_016862356.1
XM_017006868.1 1568 Intron XP_016862357.1

View Full Product Details