Product Details

SNP ID
rs201226192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108381559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGGGATGCTATTCTTCTTGAA[A/C]CTGAAAAACAGAATGTCAGTGTGTT
Phenotype
MIM: 604371 MIM: 609929
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HHLA2 PubMed Links

Gene Details

Gene
HHLA2
Gene Name
HERV-H LTR-associating 2
There are no transcripts associated with this gene.

Gene
MYH15
Gene Name
myosin heavy chain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014981.1 6005 Missense Mutation NP_055796.1
XM_011512559.2 6005 Missense Mutation XP_011510861.1
XM_017005922.1 6005 Missense Mutation XP_016861411.1

View Full Product Details