Product Details

SNP ID
rs200651923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138495057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATTTCTTTACTGCAGGTACAAT[A/G]GCATCCAAGTCATCAATTTCTGTAA
Phenotype
MIM: 614310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP70 PubMed Links

Gene Details

Gene
CEP70
Gene Name
centrosomal protein 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288964.1 2382 Silent Mutation GCC,GCT A566A NP_001275893.1
NM_001288965.1 2382 Silent Mutation GCC,GCT A564A NP_001275894.1
NM_001288966.1 2382 Intron NP_001275895.1
NM_001288967.1 2382 Silent Mutation GCC,GCT A432A NP_001275896.1
NM_001320598.1 2382 Silent Mutation GCC,GCT A584A NP_001307527.1
NM_001320599.1 2382 Intron NP_001307528.1
NM_001320600.1 2382 Intron NP_001307529.1
NM_024491.3 2382 Silent Mutation GCC,GCT A584A NP_077817.2
XM_005247805.3 2382 Intron XP_005247862.1
XM_011513209.2 2382 Intron XP_011511511.1
XM_017007270.1 2382 Intron XP_016862759.1
XM_017007271.1 2382 Intron XP_016862760.1
XM_017007272.1 2382 Intron XP_016862761.1
XM_017007273.1 2382 Intron XP_016862762.1
XM_017007274.1 2382 Intron XP_016862763.1
XM_017007275.1 2382 Silent Mutation GCC,GCT A584A XP_016862764.1
XM_017007276.1 2382 Intron XP_016862765.1
XM_017007277.1 2382 Intron XP_016862766.1
XM_017007278.1 2382 Intron XP_016862767.1
XM_017007279.1 2382 Intron XP_016862768.1
XM_017007280.1 2382 Intron XP_016862769.1
XM_017007281.1 2382 Intron XP_016862770.1
XM_017007282.1 2382 Silent Mutation GCC,GCT A325A XP_016862771.1
XM_017007283.1 2382 Silent Mutation GCC,GCT A325A XP_016862772.1
XM_017007284.1 2382 Silent Mutation GCC,GCT A325A XP_016862773.1

View Full Product Details