Product Details

SNP ID
rs199858039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40506362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGGACGCCAGCCATGTTCCAG[A/G]CCGCTTGGCGCCAGGTGAGTGAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF620 PubMed Links

Gene Details

Gene
ZNF620
Gene Name
zinc finger protein 620
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256167.1 3581 UTR 5 NP_001243096.1
NM_001256168.1 3581 UTR 5 NP_001243097.1
NM_175888.3 3581 Missense Mutation ACC,GCC T4A NP_787084.1
XM_005265011.3 3581 Missense Mutation ACC,GCC T4A XP_005265068.1
XM_005265012.3 3581 Missense Mutation ACC,GCC T4A XP_005265069.1
XM_017006064.1 3581 Missense Mutation ACC,GCC T4A XP_016861553.1
XM_017006065.1 3581 Missense Mutation ACC,GCC T4A XP_016861554.1
XM_017006066.1 3581 UTR 5 XP_016861555.1
XM_017006067.1 3581 Intron XP_016861556.1
XM_017006068.1 3581 Intron XP_016861557.1
XM_017006069.1 3581 UTR 5 XP_016861558.1

View Full Product Details