Product Details

SNP ID
rs199891591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9785325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGACACCACTCACCTGAAGGGC[C/T]TGTTCTGATTGCGAGGGGAGGTGCT
Phenotype
MIM: 604226 MIM: 601982 MIM: 602945
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC4 PubMed Links

Gene Details

Gene
ARPC4
Gene Name
actin related protein 2/3 complex subunit 4
There are no transcripts associated with this gene.

Gene
ARPC4-TTLL3
Gene Name
ARPC4-TTLL3 readthrough
There are no transcripts associated with this gene.

Gene
OGG1
Gene Name
8-oxoguanine DNA glycosylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002542.5 1734 Intron NP_002533.1
NM_016819.3 1734 Intron NP_058212.1
NM_016820.3 1734 Intron NP_058213.1
NM_016821.2 1734 Intron NP_058214.1
NM_016826.2 1734 Intron NP_058434.1
NM_016827.2 1734 Intron NP_058436.1
NM_016828.2 1734 Intron NP_058437.1
NM_016829.2 1734 Intron NP_058438.1
XM_011533760.1 1734 Intron XP_011532062.1
XM_017006493.1 1734 Intron XP_016861982.1
XM_017006494.1 1734 Intron XP_016861983.1
XM_017006495.1 1734 Intron XP_016861984.1
XM_017006496.1 1734 Intron XP_016861985.1
XM_017006497.1 1734 Intron XP_016861986.1
XM_017006498.1 1734 Intron XP_016861987.1
XM_017006499.1 1734 Intron XP_016861988.1
Gene
TADA3
Gene Name
transcriptional adaptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278270.1 1734 Missense Mutation AAG,AGG K304R NP_001265199.1
NM_006354.3 1734 Missense Mutation AAG,AGG K304R NP_006345.1
NM_133480.2 1734 Missense Mutation AAG,AGG K304R NP_597814.1

View Full Product Details