Product Details

SNP ID
rs199925649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40511559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCAGAGGGCCCTGTACAGGGA[A/G]GTGATGCTGGAGAATTATGCAAATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF620 PubMed Links

Gene Details

Gene
ZNF620
Gene Name
zinc finger protein 620
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256167.1 175 Intron NP_001243096.1
NM_001256168.1 175 Intron NP_001243097.1
NM_175888.3 175 Silent Mutation GAA,GAG E38E NP_787084.1
XM_005265011.3 175 Silent Mutation GAA,GAG E38E XP_005265068.1
XM_005265012.3 175 Silent Mutation GAA,GAG E38E XP_005265069.1
XM_017006064.1 175 Silent Mutation GAA,GAG E83E XP_016861553.1
XM_017006065.1 175 Silent Mutation GAA,GAG E38E XP_016861554.1
XM_017006066.1 175 UTR 5 XP_016861555.1
XM_017006067.1 175 UTR 5 XP_016861556.1
XM_017006068.1 175 UTR 5 XP_016861557.1
XM_017006069.1 175 Intron XP_016861558.1

View Full Product Details