Product Details
- SNP ID
-
rs200376706
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:132559006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTCAGTCTTTTGGCTTGGTCCC[A/G]CAGCTCCATTGTTGCGATTGCTGAA
- Phenotype
-
MIM: 614288
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACAD11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2270801] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACAD11
- Gene Name
- acyl-CoA dehydrogenase family member 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032169.4 |
2688 |
Missense Mutation |
CGG,TGG |
R770W |
NP_115545.3 |
- Gene
- NPHP3-ACAD11
- Gene Name
- NPHP3-ACAD11 readthrough (NMD candidate)
There are no transcripts associated with this gene.
View Full Product Details