Product Details

SNP ID
rs199659889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:94084156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCAATATGCTGTCCTGCTTAAC[C/T]GATTCAATTATCCTTTTGAACTGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSUN3 PubMed Links

Gene Details

Gene
NSUN3
Gene Name
NOP2/Sun RNA methyltransferase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022072.3 288 Nonsense Mutation CGA,TGA R58* NP_071355.1

View Full Product Details