Product Details

SNP ID
rs200823249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109819054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGCTTCATCCTTTAAAAAACT[A/G]CAGAAGGTGAGTCAAACTTATGATA
Phenotype
MIM: 606468 MIM: 605224
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAR1 PubMed Links

Gene Details

Gene
GAR1
Gene Name
GAR1 ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018983.3 730 Silent Mutation CTA,CTG L141L NP_061856.1
NM_032993.2 730 Intron NP_127460.1
Gene
RRH
Gene Name
retinal pigment epithelium-derived rhodopsin homolog
There are no transcripts associated with this gene.

View Full Product Details