Product Details
- SNP ID
-
rs200904771
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:2271341 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGCTGCGGCAGTGGTGCTTCCG[A/G]CGGATGACGGTGAAGGGTGCTTTGC
- Phenotype
-
MIM: 614176
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MXD4
PubMed Links
Gene Details
- Gene
- MXD4
- Gene Name
- MAX dimerization protein 4
There are no transcripts associated with this gene.
- Gene
- ZFYVE28
- Gene Name
- zinc finger FYVE-type containing 28
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001172656.1 |
2876 |
Silent Mutation |
CGC,CGT |
R804R |
NP_001166127.1 |
NM_001172657.1 |
2876 |
Intron |
|
|
NP_001166128.1 |
NM_001172658.1 |
2876 |
Intron |
|
|
NP_001166129.1 |
NM_001172659.1 |
2876 |
Silent Mutation |
CGC,CGT |
R764R |
NP_001166130.1 |
NM_001172660.1 |
2876 |
Intron |
|
|
NP_001166131.1 |
NM_020972.2 |
2876 |
Silent Mutation |
CGC,CGT |
R834R |
NP_066023.2 |
XM_006713900.3 |
2876 |
Silent Mutation |
CGC,CGT |
R834R |
XP_006713963.1 |
XM_006713902.3 |
2876 |
Missense Mutation |
GCC,GTC |
A908V |
XP_006713965.1 |
XM_006713904.3 |
2876 |
Intron |
|
|
XP_006713967.1 |
XM_011513540.2 |
2876 |
Silent Mutation |
CGC,CGT |
R720R |
XP_011511842.1 |
XM_011513541.2 |
2876 |
Intron |
|
|
XP_011511843.1 |
XM_017008502.1 |
2876 |
Silent Mutation |
CGC,CGT |
R776R |
XP_016863991.1 |
XM_017008503.1 |
2876 |
Missense Mutation |
GCC,GTC |
A909V |
XP_016863992.1 |
XM_017008504.1 |
2876 |
Missense Mutation |
GCC,GTC |
A909V |
XP_016863993.1 |
XM_017008505.1 |
2876 |
Silent Mutation |
CGC,CGT |
R720R |
XP_016863994.1 |
XM_017008506.1 |
2876 |
Intron |
|
|
XP_016863995.1 |
XM_017008507.1 |
2876 |
Intron |
|
|
XP_016863996.1 |
XM_017008508.1 |
2876 |
Intron |
|
|
XP_016863997.1 |
XM_017008509.1 |
2876 |
Intron |
|
|
XP_016863998.1 |
XM_017008510.1 |
2876 |
Intron |
|
|
XP_016863999.1 |
View Full Product Details