Product Details
- SNP ID
-
rs200294648
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:5525430 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTGAATTTGGGAAGGAAGGAACA[C/T]GCAAAAATTTTTACCTTCTTCTTTC
- Phenotype
-
MIM: 607261
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EVC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs886532] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EVC2
- Gene Name
- EvC ciliary complex subunit 2
There are no transcripts associated with this gene.
- Gene
- LINC01587
- Gene Name
- long intergenic non-protein coding RNA 1587
There are no transcripts associated with this gene.
View Full Product Details