Product Details

SNP ID
rs200347007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109689395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAGGCAAAACAGGGAACCTGCT[G/T]CTTTCCAATTGCACTTGGGTCTTTG
Phenotype
MIM: 601532
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CASP6 PubMed Links

Gene Details

Gene
CASP6
Gene Name
caspase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001226.3 628 Missense Mutation AAG,CAG K273Q NP_001217.2
NM_032992.2 628 Missense Mutation AAG,CAG K184Q NP_116787.1
Gene
MCUB
Gene Name
mitochondrial calcium uniporter dominant negative beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017918.4 628 Intron NP_060388.2
XM_006714246.3 628 Intron XP_006714309.1

View Full Product Details