Product Details
- SNP ID
-
rs200382055
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:73869478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCGCCTGCTGAGCCCCATGGCCC[G/T]CGCTGCTCTCTCCGCCGCCCCCAGC
- Phenotype
-
MIM: 155730
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CXCL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11547681] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CXCL1
- Gene Name
- C-X-C motif chemokine ligand 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001511.3 |
87 |
Missense Mutation |
CGC,CTC |
R3L |
NP_001502.1 |
View Full Product Details