Product Details

SNP ID
rs200734239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:77160944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAACTTTTTGCACTTATACCATA[C/G]TATTATACTTTGTCATACTTCAGAA
Phenotype
MIM: 603203
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCNG2 PubMed Links

Gene Details

Gene
CCNG2
Gene Name
cyclin G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004354.2 943 Missense Mutation ACT,AGT T167S NP_004345.1
XM_011532398.1 943 Missense Mutation ACT,AGT T167S XP_011530700.1
XM_011532399.2 943 Missense Mutation ACT,AGT T167S XP_011530701.1

View Full Product Details