Product Details

SNP ID
rs199476186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186194538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCATTGAGTACACAGAGGAATAC[C/T]GCCACATGCCGCTGCTGAAGCTCTG
Phenotype
MIM: 608614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP4V2 PubMed Links

Gene Details

Gene
CYP4V2
Gene Name
cytochrome P450 family 4 subfamily V member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207352.3 342 Missense Mutation CGC,TGC R85C NP_997235.3
XM_005262935.3 342 Missense Mutation CGC,TGC R85C XP_005262992.1
XM_017008037.1 342 UTR 5 XP_016863526.1
Gene
FLJ38576
Gene Name
uncharacterized LOC651430
There are no transcripts associated with this gene.

View Full Product Details