Product Details

SNP ID
rs199867627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99905618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACGATCTGGGATATAAGGAATAA[A/G]GAGGATTAGAGACCATCAACTGGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB14 PubMed Links

Gene Details

Gene
DNAJB14
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031723.3 1360 Missense Mutation CCT,CTT P274L NP_001026893.1
NM_001278310.1 1360 Missense Mutation CCT,CTT P207L NP_001265239.1
NM_001278311.1 1360 Intron NP_001265240.1
XM_011532262.2 1360 Missense Mutation CCT,CTT P189L XP_011530564.1
XM_011532263.2 1360 Missense Mutation CCT,CTT P189L XP_011530565.1
XM_011532265.2 1360 Missense Mutation CCT,CTT P186L XP_011530567.1
XM_017008628.1 1360 Missense Mutation CCT,CTT P186L XP_016864117.1

View Full Product Details